Anti-CD83 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HB15e]
Catalog # 75843-920
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Name:CD83
- Antigen Symbol:CD83
- Clonality:Monoclonal
- Clone:HB15e
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Cat. No.:75843-920
- Supplier no.:05911-50-100
Specifications
About this item
The HB15e monoclonal antibody specifically reacts with human CD83, a 43kDA single chain type I glycoprotein also known as HB15. A useful marker for mature human dendritic cells, it is also expressed on some activated lymphocytes. CD83’s precise role has not been deduced, but it is thought to be involved in antigen presentation.
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human