Anti-CD73 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AD2]
Catalog # 75843-916
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD73
- Clonality:Monoclonal
- Clone:AD2
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Antigen Synonyms:Ecto-5'-nucleotidase|NT5E
- Cat. No.:75843-916
- Supplier no.:05811-80-100
Specifications
About this item
The AD2 monoclonal antibody specifically reacts with human CD73 (ecto-5’-nucleotidase), a 70 kDA glycosyl phosphatidylinositol (GPI)-anchored glycoprotein. CD73 catalyzes the dephosphorylation of adenosine monophosphate (AMP) to adenosine and is expressed on subsets of B, T, dendritic, endothelial, and mesenchymal stem cells. It is also involved in the adhesion of lymphocytes to endothelium and T cell activation.
Type: Primary
Antigen: CD73
Clonality: Monoclonal
Clone: AD2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human