Anti-CD62L Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DREG-56]
Catalog # 75841-650
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Name:L-selectin
- Antigen Symbol:CD62L
- Clonality:Monoclonal
- Clone:DREG-56
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Antigen Synonyms:LAM-1|MEL-14|Ly-22|LECAM-1|L-selectin
- Cat. No.:75841-650
- Supplier no.:04731-60-100
Specifications
About this item
The DREG-56 monoclonal antibody specifically reacts with human CD62L, also known as L-selectin and LECAM-1. It mediates the migration of lymphocytes to the site of inflammation and their return to the peripheral lymphoid tissues and to the HEV (high endothelial venules). CD62L is expressed on monocytes, NK cells, neutrophils, granulocytes, and subsets of T and B cells. It is reported that the DREG-56 antibody inhibits the binding of lymphocytes to HEV tissue in frozen peripheral tissue.
Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: DREG-56
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human