Anti-CD34 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4H11]
Catalog # 75841-796
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD34
- Clonality:Monoclonal
- Clone:4H11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:gp 105-120|Class III epitope|Mucosialin|My10
- Cat. No.:75841-796
- Supplier no.:06411-50-25
Specifications
About this item
The 4H11 monoclonal antibody specifically reacts with the Class III epitope of CD34 (mucosialin), a transmembrane phosphoglycoprotein. It is expressed on most hematopoietic progenitors, pluripotent stem cells and tumor cells.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human