Specifications
- Antibody Type:Primary
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:145-2C11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Size:25 μg
- Form:liquid
- Antigen Synonyms:T3|CD3 epsilon
- Cat. No.:75841-728
- Supplier no.:05122-50-25
Specifications
About this item
The 145-2C11 monoclonal antibody reacts with mouse CD3e, the 20 kDa ε chain of the TCR complex. Together with the γ and δ subunits of CD3, the ε subunits are involved in the assembly, trafficking, and surface expression of T-cell receptor complex. CD3 is expressed on thymocytes, mature T cells, and natural killer T cells, and the ε chain enhances the antigen recognition.The 145-2C11 antibody binds to the TCR complex and, depending on the conditions, initiates T cells activation, proliferation, and apoptosis. The soluble antibody seems to block lysis of target cells by antigen-specific cytotoxic T lymphocytes. The 145-2C11 antibody does not cross-react with the rat leukocytes, and it is used as a phenotypic marker for mouse T lymphocytes.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse