Anti-CD3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: G4.18]
Catalog # 75841-694
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:G4.18
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Reactivity:Rat
- Size:50 μg
- Form:liquid
- Antigen Synonyms:T3
- Cat. No.:75841-694
- Supplier no.:05113-50-50
Specifications
About this item
The G4.18 monoclonal antibody specifically reacts with rat CD3, a T cell lineage marker and associated with the T cell receptor (TCR). CD3 is involved in antigen recognition, cell activation, and signal transduction and is found on subsets of NK-T, peripheral T, dendritic epidermal T, and thymocyte cells. Immobilized G4.18 antibody activates T cells in vitro and in vivo treatments of the G4.18 antibody prevents cardiac and skin allograft rejection.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: G4.18
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Rat