Anti-CD27 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: O323]
Catalog # 75843-048
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Name:CD27 molecule
- Antigen Symbol:CD27
- Clonality:Monoclonal
- Clone:O323
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Antigen Synonyms:Tp55|S152|T14|TNFRSF7|S152. LPFS2
- Cat. No.:75843-048
- Supplier no.:09311-50-100
Specifications
About this item
The O323 monoclonal antibody specifically binds to the human CD27, which is a lymphocyte-specific member of the TNF/NGF-R family, expressed on a subset of human thymocytes and the majority of the mature T cells. CD27 is upregulated by T lymphocytes stimulation, and binds to CD70, enhancing the interaction between T and B lymphocytes. CD27 is a marker of memory B cells.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human