Anti-CD144 (VE-Cadherin) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 16B1]
Catalog # 75843-146
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Name:CD144 (VE-Cadherin)
- Antigen Symbol:CD144 (VE-Cadherin)
- Clonality:Monoclonal
- Clone:16B1
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Form:liquid
- Antigen Synonyms:Cadherin-5|VE-Cadherin
- Cat. No.:75843-146
- Supplier no.:16511-60-100
Specifications
About this item
The 16B1 monoclonal antibody specifically reacts with human CD144, the 140 kDA molecule called vascular endothelial (VE-cadherin) or cadherin 5. CD144 is an endothelial specific calcium-dependent adhesion molecule involved in cell contact-dependent growth inhibition, migration, survival, adhesion. It is concentrated at the intercellular boundaries of endothelial cells and essential in maintaining cell layer integrity.
Type: Primary
Antigen: CD144 (VE-Cadherin)
Clonality: Monoclonal
Clone: 16B1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human