- Antibody Type:Primary
- Antigen Name:Integrin, alpha X
- Antigen Symbol:CD11c
- Clonality:Monoclonal
- Clone:N418
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow Cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Antigen Synonyms:ITGAX|CR4|p150
- Cat. No.:75841-550
- Supplier no.:03212-87-100
The N418 monoclonal antibody specifically reacts with the integrin αx chain of the mouse CD11c, which is expressed on dendritic cells, CD4-/CD8+ intestinal intraepithelial lymphocytes (IEL) and some activated T lymphocytes. Low levels of CD11c were detected on mouse splenic natural killer cells and on the monocyte/macrophage lineage cells.CD11c expression is upregulated on IEL and T lymphocytes after activation. It binds to CD54, fibrinogen and iC3b and influences the leukocyte adhesive interactions. The N418 antibody binds to CD11c on splenic dendritic cells of the mouse in the T-dependent areas. It also contributes to the binding of iC3b.
Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse