Anti-CD11b (ICRF44) Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: ICRF44]
Catalog # 75841-510
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD11b (ICRF44)
- Clonality:Monoclonal
- Clone:ICRF44
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:C3biR|Mo1|CR3|Mac-1|ITGAM
- Cat. No.:75841-510
- Supplier no.:03211-77-25
Specifications
About this item
The ICRF44 monoclonal antibody specifically reacts with the 165 kDa human adhesion glycoprotein CD11b, which forms, together with the 95 kDa CD18 (integrin β2) a complex known as Mac-1. CD11b is expressed on the surface of activated lymphocytes, a subset of natural killer cells, granulocytes, and monocytes. It functions as a receptor in cell-cell and cell-matrix interactions and as a receptor for iC3b, ICAM-1, ICAM-2, and ICAM-3 intercellular adhesion molecules.
Type: Primary
Antigen: CD11b (ICRF44)
Clonality: Monoclonal
Clone: ICRF44
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human