Anti-CCR7 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3D12]
Catalog # 75843-168
Supplier: Biogems
Specifications
- Antibody Type:Primary
- Antigen Symbol:CCR7
- Clonality:Monoclonal
- Clone:3D12
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Antigen Synonyms:EBI-1|MIP-3 beta Receptor
- Cat. No.:75843-168
- Supplier no.:20011-50-100
Specifications
About this item
The 3D12 monoclonal antibody specifically reacts with human CD197 (CCR7 or EBI-1), a seven-transmembrane G-protein-coupled. It is the receptor for the secondary lymphoid-tissue chemokine (SLC or 6CKine), CCL19, and CCL21. CD197 is expressed on subsets of B and T cells, and mature dendritic cells. The monoclonal 3D12 antibody recognizes the N-terminus of the receptor.
We recommend that this antibody be incubated for at least 45 minutes at 4°C for optimal staining.
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 3D12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human