Order Entry
United States
ContactUsLinkComponent
Human Recombinant MMP-10 (from NS0 Cells)
Human Recombinant MMP-10 (from NS0 Cells)
Catalog # 103010-388
Supplier:  Anaspec Inc
CAS Number:  
Human Recombinant MMP-10 (from NS0 Cells)
Catalog # 103010-388
Supplier:  Anaspec Inc
Supplier Number:  AS-72067
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Human
  • Size:
    100 µl
  • Storage Conditions:
    –78 °C
  • Protein Synonyms:
    MMP-10 recombinant|MMP-10|Matrix metalloproteinase 10
  • UniProtKB:
    P09238
  • Protein/Peptide Name:
    MMP-10
  • Molecular Weight:
    58 kDa
  • Concentration:
    10 μg/mL
  • Cat. No.:
    103010-388
  • Supplier no.:
    AS-72067

Specifications

About this item

Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP-10 (stromelysin 2) is involved in several pathological conditions, such as cancer, arthritis and wound healing. MMP-10 is secreted as zymogen with a prodomain, a catalytic domain, a hinge region, and a hemopexin-like domain. It can activate other MMPs such as MMP-1, MMP-8 and degrade a variety of substrates, including gelatin, collagens III, IV and V, fibronectin, aggrecan

This recombinant human MMP-10 was expressed as a pro-enzyme enzyme from its DNA sequence7 transfected into a mouse myeloma cell line, NS0. The apparent Mr on SDS-PAGE is 58-kDa. Incubation with 1 mM APMA at 37°C for 1-2 hours will activate pro-MMP-10. Its activity can be measured by FRET peptides