Order Entry
United States
ContactUsLinkComponent
Adhesive Vinyl Labels 'Triple Rinsed', Labelmaster
Adhesive Vinyl Labels 'Triple Rinsed', Labelmaster
Catalog # 101378-234
Supplier:  Label Master
Adhesive Vinyl Labels 'Triple Rinsed', Labelmaster
Catalog # 101378-234
Supplier:  Label Master
Supplier Number:  TR333

Specifications

  • Description:
    Preprinted adhesive labels
  • Dimensions:
    152.4×152.4 mm (6×6")
  • Material:
    Semi-Gloss Paper
  • Cat. No.:
    101378-234
  • Legend:
    Triple Rinsed
  • Supplier no.:
    TR333

Specifications

About this item

These triple rinsed labels are used to identify containers that have been triple rinsed in accordance with EPA regulations in 40 CFR part 261.7.

  • Constructed of paper
  • Offer an economical alternative to vinyl

Color: White, blue and black