Specifications
- Antibody Type:Primary
- Antigen Name:ADP-ribosylation factor 6
- Antigen Symbol:ARF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:P62330
- Molecular Weight:20082
- Storage Temperature:At −20˚C for one year. After reconstitution, at 4 ˚C for one month. It can also be aliquotted and stored frozen at −20 ˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2 ml of distilled water will yield a concentration of 500 μg/ml
- Shipping Temperature:4 °C
- Immunogen:E.coli-derived human ARF6 recombinant protein (Position: G2-S175). Human ARF6 shares 100% amino acid (aa) sequence identity with both mouse and rat ARF6.
- Purification:Immunogen affinity purified
- Cat. No.:76174-788
Specifications
About this item
Rabbit IgG polyclonal antibody for ADP-ribosylation factor 6(ARF6) detection. Tested with WB in Human;Mouse;Rat.
1. "Entrez Gene: ARF6 ADP-ribosylation factor 6".
2. Prigent M, Dubois T, Raposo G, Derrien V, Tenza D, Rossé C, Camonis J, Chavrier P (December 2003). "ARF6 controls post-endocytic recycling through its downstream exocyst complex effector". J. Cell Biol. 163(5): 1111–21.
3. Peru y Colón de Portugal RL, Acevedo SF, Rodan AR, Chang LY, Eaton BA, Rothenfluh A (December 2012)."Adult neuronal Arf6 controls ethanol-induced behavior with Arfaptin downstream of Rac1 and RhoGAP18B".J. Neurosci. 32 (49): 17706–13.
Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ARF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat