Anti-TK1 IgG Polyclonal Antibody
76174-520
- Antibody Type:Primary
- Antigen Symbol:TK1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:P04183
- Molecular Weight:25469
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:E. coli-derived human TK1 recombinant protein (Position: S2-N234). Human TK1 shares 87.1% and 91.7% amino acid (aa) sequence identity with mouse and rat TK1, respectively.
- Purification:Immunogen affinity purified.
- Cat. No.:76174-520
Rabbit IgG polyclonal antibody for Thymidine kinase, cytosolic(TK1) detection. Tested with WB in Human.
Type: Primary
Antigen: TK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human