Anti-CCL5 IgG Polyclonal Antibody
Catalog # 76174-002
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Symbol:CCL5
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:P13501
- Molecular Weight:9990
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:E. coli-derived human RANTES recombinant protein (Position: Y26-S91). Human RANTES shares 83.3% amino acid (aa) sequence identity with both mouse and rat RANTES.
- Purification:Immunogen affinity purified.
- Cat. No.:76174-002
Specifications
About this item
Rabbit IgG polyclonal antibody for C-C motif chemokine 5(CCL5) detection. Tested with WB, ELISA in Human.
Type: Primary
Antigen: CCL5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human