- Antibody Type:Primary
- Antigen Name:NLR family, CARD domain containing 4
- Antigen Symbol:NLRC4
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:Q9NPP4
- Antigen Synonyms:CLANA|AIFEC|CLAN|FCAS4|CARD12|CLAN1|CLR2.1|CLANC|IPAF|CLAND|CLANB
- UniProtKB:Q9NPP4
- Molecular Weight:116159
- Storage Temperature:At −20 °C for one year. After reconstitution, at 4 °C for one month. It can also be aliquotted and stored frozen at −20 °C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human CARD12 (838-874aa KILAQNLHNLVKLSILDLSENYLEKDGNEALHELIDR), different from the related mouse sequence by five amino acids, and from the related rat sequence by seven amino acids.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-810
Rabbit IgG polyclonal antibody for NLR family CARD domain-containing protein 4(NLRC4) detection. Tested with WB in Human;Mouse;Rat.
NLR family CARD domain-containing protein 4, also known as CARD12, is a protein that in humans is encoded by the NLRC4 gene. It is mapped to 2p22.3. This gene encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
Type: Primary
Antigen: NLRC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat