Specifications
- Antibody Type:Primary
- Antigen Name:Serine/arginine-rich Splicing Factor 1
- Antigen Symbol:SRSF1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized
- Gene ID:6426
- Antigen Synonyms:ASF|SF2|SRp30a|SFRS1|SF2p33
- UniProtKB:Q07955
- Molecular Weight:27745
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of Human SF2(6-33aa VIRGPAGNNDCRIYVGNLPPDIRTKDIE), identical to the related mouse sequence.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-642
- Packaging:Vial
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine/arginine-rich splicing factor 1(SRSF1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
SF2, also known as Serine/arginine-rich splicing factor 1 (SRSF1), is a protein that in humans is encoded by the SFRS1 gene. This gene encodes a member of the arginine/serine-rich splicing factor protein family. There is a pseudogene of this gene on chromosome 13. The encoded protein can either activate or repress splicing, depending on its phosphorylation state and its interaction partners. Multiple transcript variants have been found for this gene. ASF/SF2 is necessary for all splicing reactions to occur, and influences splice site selection in a concentration-dependent manner, resulting in alternative splicing.
Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.
Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.
WB: The detection limit for SF2 is approximately 0.2 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: STAT6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
, Human