Specifications
- Antibody Type:Primary
- Antigen Name:Rho-associated, coiled-coil containing protein kinase 2
- Antigen Symbol:ROCK2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized
- Gene ID:9475
- Antigen Synonyms:ROCK-II
- UniProtKB:O75116
- Molecular Weight:160900
- Storage Temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4°C
- Immunogen:E.coli-derived human ROCK2 recombinant protein (Position: E652-D923). Human ROCK2 shares 94.9% and 95.2% amino acid (aa) sequence identity with mouse and rat ROCK2, respectively.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-608
- Packaging:Vial
Specifications
About this item
Rabbit IgG polyclonal antibody for Rho-associated protein kinase 2(ROCK2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Rho-associated kinase (ROCK), including the ROCK-I and ROCK-II isoforms, is a protein kinase involved in signaling from Rho to actin cytoskeleton. Serine/threonine kinase ROCK II/Rho kinase, which is an isozyme of ROCK I, is one of the targets for the small GTPase Rho. ROCK II regulates the formation of actin stress fibers and focal adhesions, cytokinesis, smooth muscle contraction, and the activation of c-fos serum response element. Sequencing analysis has shown that human ROCK II contains 1388 amino acid residues with a calculated molecular mass of approximately 161 kDa. Fluorescence in situ hybridization analysis showed that the human ROCK II gene is located on chromosome 2p24. Thumkeo et al. concluded that ROCK-II is essential in inhibiting blood coagulation and maintaining blood flow in the endothelium-free labyrinth layer and that loss of ROCK-II leads to thrombus formation, placental dysfunction, intrauterine growth retardation, and fetal death.
Add 0.2 ml of distilled water will yield a concentration of 500µg/ ml.
Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.
WB: The detection limit for ROCK2 is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: SDC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat