Specifications
- Antibody Type:Primary
- Antigen Symbol:SERPINB2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:P05120
- Molecular Weight:46596
- Storage Temperature:At −20 ˚C for one year. After reconstitution, at 4 ˚C for one month. It can also be aliquotted and stored frozen at −20 ˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2 ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4 °C
- Immunogen:E.coli-derived human SerpinB2 recombinant protein (Position: M1-K180). Human SerpinB2 shares 70% and 66% amino acid (aa) sequence identity with mouse and rat SerpinB2, respectively.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-544
Specifications
About this item
Rabbit IgG polyclonal antibody for Plasminogen activator inhibitor 2(SERPINB2) detection. Tested with WB, IHC-P in Human.
SERPINB2 is also known as PAI or PAI2. Plasminogen activator inhibitor-2 (placental PAI) is acoagulation factor that inactivates tPA and urokinase. It is present in most cells, especially monocytes/macrophages. PAI-2 exists in two forms, a 60-kDa extracellular glycosylated form and a 43-kDa intracellular form. It is present only at detectable quantities in blood during pregnancy, as it is produced by the placenta, and may explain partially the increased rate of thrombosisduring pregnancy. The majority of expressed PAI-2 remains unsecreted due to the presence of an inefficient internal signal peptide.
Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.
Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05 mg NaN3.
WB: The detection limit for SerpinB2 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: SFRP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Yes
Host: Rabbit
Isotype: IgG
Reactivity: Human