Specifications
- Antibody Type:Primary
- Antigen Symbol:APOE
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg/vial
- Form:Lyophilized
- Gene ID:P02649
- Molecular Weight:36154
- Storage Temperature:At −20 ˚C for one year. After reconstitution, at 4 ˚C for one month. It can also be aliquotted and stored frozen at −20 ˚C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2 ml of distilled water will yield a concentration of 500 μg/ml.
- Shipping Temperature:4 °C
- Immunogen:E.coli-derived human Apolipoprotein E recombinant protein (Position: K19-H317). Human Apolipoprotein E shares 73% and 72% amino acid (aa) sequence identity with mouse and rat Apolipoprotein E, respectively.
- Purification:Immunogen affinity purified.
- Cat. No.:76173-488
Specifications
About this item
Rabbit IgG polyclonal antibody for Apolipoprotein E(APOE) detection. Tested with WB, IHC-P, ELISA(Cap) in Human.
APOE is also known as AD2 or LPG. The protein encoded by this gene is a major apoprotein of the chylomicron. It binds to a specific liver and peripheral cell receptor, and is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. This gene maps to chromosome 19 in a cluster with the related apolipoprotein C1 and C2 genes. Mutations in this gene result in familial dysbetalipoproteinemia, or type III hyperlipoproteinemia (HLP III), in which increased plasma cholesterol and triglycerides are the consequence of impaired clearance of chylomicron and VLDL remnants. Alternative splicing results in multiple transcript variants.
Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.
Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05 mg NaN3.
WB: The detection limit for Apolipoprotein E is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: ATP5H
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat