Order Entry
United States
ContactUsLinkComponent
Anti-MED4 IgG Polyclonal Antibody
Anti-MED4 IgG Polyclonal Antibody
Catalog # 76171-968
Anti-MED4 IgG Polyclonal Antibody
Catalog # 76171-968
Supplier Number:  A06467-1

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    MED4
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg/vial
  • Form:
    Lyophilized
  • Gene ID:
    MED4: Q9NPJ6
  • Storage Temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Concentration:
    Add 0.2ml of distilled water will yield a concentration of 500 μg/ml.
  • Shipping Temperature:
    4°C
  • Immunogen:
    E. coli-derived human MED4 recombinant protein (Position: R27-K140).
  • Purification:
    Immunogen affinity purified.
  • Cat. No.:
    76171-968

Specifications

About this item

Rabbit IgG polyclonal antibody for MED4 detection. Tested with WB, IHC-P, Direct ELISA in Human;Mouse;Rat.

Type: Primary
Antigen: MED4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat