Human OSM/Oncostatin M EZ-Set; ELISA Kit (DIY Antibody Pairs), Boster
Catalog # 76172-808
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:OSM
- Description:Human OSM/Oncostatin M EZ-Set; ELISA Kit (DIY Antibody Pairs)
- Size:For 5 plates, 96 wells each
- Cat. No.:76172-808
Specifications
About this item
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects OSM/Oncostatin M in Human samples
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects OSM/Oncostatin M in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.
The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.