Mouse PON1/Paraoxonase 1 PicoKine; ELISA Kit, Boster
Catalog # 76172-782
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse PON1/Paraoxonase 1 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-782
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse PON1
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Mouse Paraoxonase 1/Pon1 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Paraoxonase 1/Pon1 tissue specificity: Plasma, liver, kidney, heart, brain, small intestine and lung. In the plasma, associated with HDL.
96wells/kit, with removable strips.