Order Entry
United States
ContactUsLinkComponent
Human Siglec5/Cd170 PicoKine; ELISA Kit, Boster
Human Siglec5/Cd170 PicoKine; ELISA Kit, Boster
Catalog # 76172-742
CAS Number:  
undefined
Human Siglec5/Cd170 PicoKine; ELISA Kit, Boster
Catalog # 76172-742
Supplier Number:  EK1583
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Human Siglec5/Cd170 PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-742

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human Siglec5

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Human Cd170/SIGLEC5 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cd170/SIGLEC5 tissue specificity: Expressed by monocytic/myeloid lineage cells. Found at high levels in peripheral blood leukocytes, spleen, bone marrow and at lower levels in lymph node, lung, appendix, placenta, pancreas and thymus. Expressed by monocytes and neutrophils but absent from leukemic cell lines representing early stages of myelomonocytic differentiation.

96wells/kit, with removable strips.