Rat ALCAM/Cd166 PicoKine; ELISA Kit, Boster
76172-732
:
- Assay Type:Sandwich
- Description:Rat ALCAM/Cd166 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-732
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Rat ALCAM
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Rat Cd166/Alcam with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cd166/Alcam tissue specificity: Strongest expression in the lung, then brain, liver, and kidney. Present in the somatosensory system, basal ganglia, cortex, olfactory system, and circumventricular organs.
96wells/kit, with removable strips.