Order Entry
United States
ContactUsLinkComponent
Mouse Meteorin/METRN PicoKine; ELISA Kit, Boster
Mouse Meteorin/METRN PicoKine; ELISA Kit, Boster
Catalog # 76172-696
CAS Number:  
undefined
Mouse Meteorin/METRN PicoKine; ELISA Kit, Boster
Catalog # 76172-696
Supplier Number:  EK1551
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Meteorin/METRN PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-696

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse Meteorin/METRN

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Mouse Meteorin/Metrn with <12pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Meteorin/Metrn tissue specificity: Highly expressed in brain. Expressed in undifferentiated neural progenitors and in astrocyte lineage, particulary in Bergmann glia, a subtype of radial glia, and a few discrete neuronal populations residing in the superior colliculus, the ocular motor nucleus, the raphe and pontine nuclei, and in various thalamic nuclei. Weakly expressed in heart, kidney, skeletal muscle, spleen, testis, gut and lung. .

96wells/kit, with removable strips.