Order Entry
United States
ContactUsLinkComponent
Human APOA1/Apoa I PicoKine; ELISA Kit, Boster
Human APOA1/Apoa I PicoKine; ELISA Kit, Boster
Catalog # 76172-556
CAS Number:  
undefined
Human APOA1/Apoa I PicoKine; ELISA Kit, Boster
Catalog # 76172-556
Supplier Number:  EK1456
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Description:
    Human APOA1/Apoa I PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-556

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human APOA1

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Human Apoa I/APOA1 with <50pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(heparin, EDTA) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Apoa I/APOA1 tissue specificity: Major protein of plasma HDL, also found in chylomicrons. Synthesized in the liver and small intestine. The oxidized form at Met-110 and Met-136 is increased in individuals with increased risk for coronary artery disease, such as in carrier of the eNOSa/b genotype and exposure to cigarette smoking. It is also present in increased levels in aortic lesions relative to native ApoA-I and increased levels are seen with increasing severity of disease. .

96wells/kit, with removable strips.