Mouse CCL25/TECK PicoKine; ELISA Kit, Boster
76172-528
:
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse CCL25/TECK PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-528
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse CCL25/TECK
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Mouse Teck/Ccl25 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Teck/Ccl25 tissue specificity: Specifically expressed by thymic dendritic cells. High levels in thymus and small intestine.
96wells/kit, with removable strips.