Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse ALCAM/Cd166 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-398
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Mouse ALCAM
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Mouse Cd166/Alcam with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Cd166/Alcam tissue specificity: Detected on brain motor neurons, in differentiating retinal ganglion cells and in adult retina (PubMed:15345243). Detected on leukocytes and on lymphatic endothelial cells (PubMed:23169771). Detected in spleen B cells and T-cells (at protein level) (PubMed:9209500). Detected in adult brain and embryonic spinal cord (PubMed:15345243). Expressed at high levels in the brain, and lung, and at lower levels in the liver, and the kidney, as well as by activated leukocytes (PubMed:9209500). .
96wells/kit, with removable strips.