Chinese Hamster Endothelin PicoKine; ELISA Kit, Boster Biological Technology
Catalog # 76172-274
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Description:Chinese Hamster Endothelin PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-274
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Chinese hamster Endothelin
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Chinese Endothelin 1/EDN1 with <0.5pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Endothelin 1/EDN1 tissue specificity: Expressed in lung, placental stem villi vessels and in cultured placental vascular smooth muscle cells. .
96wells/kit, with removable strips.