Pig porcine Cadherin-2/N-Cadherin PicoKine; ELISA Kit, Boster Biological Technology
Catalog # 76172-186
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Conjugate ELISA:Avidin-Biotin-Peroxidase
- Format:Pre-coated
- Primary antibody reactivity:Pig
- Target protein:N-Cadherin-2 (CDH2)
- Description:Pig porcine Cadherin-2/N-Cadherin PicoKine; ELISA Kit
- Size:96 wells/kit
- Sample Type:Cell culture supernatants, serum and plasma (heparin, EDTA)
- Detection Range:156 - 10,000 pg/ml
- Sensitivity:<20 pg/ml
- Immunogen:Sequence: D160-A724
- Cat. No.:76172-186
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Pig porcine Cadherin-2/N-Cadherin
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Pig N-Cadherin/CDH2 with <20pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. N-Cadherin/CDH2 tissue specificity:
96wells/kit, with removable strips.