Order Entry
United States
ContactUsLinkComponent
Rabbit TGF Beta 1 PicoKine; ELISA Kit, Boster
Rabbit TGF Beta 1 PicoKine; ELISA Kit, Boster
Catalog # 76172-152
CAS Number:  
undefined
Rabbit TGF Beta 1 PicoKine; ELISA Kit, Boster
Catalog # 76172-152
Supplier Number:  EK0513-RB
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Rabbit
  • Target protein:
    TGF beta 1
  • Description:
    Rabbit TGF Beta 1 PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-152

Specifications

About this item

Sandwich High Sensitivity ELISA kit for Quantitative Detection of activated rabbit TGF beta 1

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Rabbit Tgf Beta 1/TGFB1 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(EDTA) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tgf Beta 1/TGFB1 tissue specificity: Highly expressed in bone. Abundantly expressed in articular cartilage and chondrocytes and is increased in osteoarthritis (OA). Colocalizes with ASPN in chondrocytes within OA lesions of articular cartilage. .

96wells/kit, with removable strips.