Monkey primate TGF Beta 1 PicoKine; ELISA Kit, Boster
76172-150
:
- Assay Type:Sandwich
- Primary antibody reactivity:Monkey
- Target protein:TGF beta 1
- Description:Monkey primate TGF Beta 1 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-150
Sandwich High Sensitivity ELISA kit for Quantitative Detection of activated monkey primate TGF beta 1
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Monkey Tgf Beta 1/TGFB1 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(EDTA) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tgf Beta 1/TGFB1 tissue specificity: Highly expressed in bone. Abundantly expressed in articular cartilage and chondrocytes and is increased in osteoarthritis (OA). Colocalizes with ASPN in chondrocytes within OA lesions of articular cartilage. .
96wells/kit, with removable strips.