Order Entry
United States
ContactUsLinkComponent
 
Dog Canine TGF Beta 1 PicoKine; ELISA Kit, Boster Biological Technology
  76172-146
 :  
undefined
Dog Canine TGF Beta 1 PicoKine; ELISA Kit, Boster Biological Technology
  76172-146
 :  EK0513-CN
 :  

 

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Dog
  • Target protein:
    TGF beta 1
  • Description:
    Dog Canine TGF Beta 1 PicoKine; ELISA Kit
  • Size:
    96 wells/kit
  • Cat. No.:
    76172-146

 

 

Sandwich High Sensitivity ELISA kit for Quantitative Detection of activated Dog Caninea TGF beta 1

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Dog Tgf Beta 1/TGFB1 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma(EDTA) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tgf Beta 1/TGFB1 tissue specificity: Highly expressed in bone. Abundantly expressed in articular cartilage and chondrocytes and is increased in osteoarthritis (OA). Colocalizes with ASPN in chondrocytes within OA lesions of articular cartilage. .

96wells/kit, with removable strips.