Horse Equine BMP-5 PicoKine; ELISA Kit, Boster
Catalog # 76172-098
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Description:Horse equine BMP-5 PicoKine; ELISA Kit
- Size:96 wells/kit
- Cat. No.:76172-098
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of horse equine BMP-5
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Horse BMP5 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and bone tissue. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. BMP5 tissue specificity: Expressed in the lung and liver.
96wells/kit, with removable strips.