Order Entry
United States
ContactUsLinkComponent
Anti-RUNX1/AML1 Polyclonal Antibody
Anti-RUNX1/AML1 Polyclonal Antibody
Catalog # 76173-192
undefined
Anti-RUNX1/AML1 Polyclonal Antibody
Catalog # 76173-192
Supplier Number:  PB9157

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    RUNX1/AML1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Cat. No.:
    76173-192

Specifications

About this item

Polyclonal antibody for CBFA2/RUNX1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CBFA2/RUNX1 information: Molecular Weight: 48737 MW; Subcellular Localization: Nucleus; Tissue Specificity: Expressed in all tissues examined except brain and heart. Highest levels in thymus, bone marrow and peripheral blood.

Type: Primary
Antigen: RUNX1/AML1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat