About this item
Gravity convection provides gentle air flow and minimal drying out.
- Intuitive user interface for setting temperature
- Large, easy to read vacuum fluorescent display
- Internal glass door allows sample viewing without impacting temperature
- Exceptionally small footprint
- Easy to clean, corrosion-resistant stainless-steel interior (AISI 430)
The gravity convection incubator provides temperatures at or below ambient: +5 to 75°C. It has a temperature uniformity of ±0.6 °C (at 37 °C) and temperature stability of ± 0.2 °C (at 37 °C).