ContactUsLinkComponent
Avantor® Alltima, Phenyl HPLC Columns
Avantor® Alltima, Phenyl HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® Alltima, Phenyl HPLC Columns
Supplier:  Avantor
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 75797-302
  • 75797-300
  • 75797-304
  • 75797-288
  • 75797-298
  • 76398-446
  • 75830-850
  • 75830-372
  • Description:
  • 75797-304
  • All-Guard cartridge for 3.0 – 4.6 mm ID columns (Use with holder 80101/N and coupler HI-081)
  • All-Guard cartridge for 2.1 mm ID columns (Use with holder 80101/N and coupler HI-081)
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon Load:
  • 7.5%
  • Endcapped:
  • Yes
  • Surface Area:
  • 340 m2/g
  • USP Code:
  • L11
  • Environmental Certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)