Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:86088-83-7
- Source:Human plasma
- Species:Human
- Size:1 mg
- Storage Conditions:Store at –20 °C.
- Protein/Peptide Name:Fibronectin
- Purity:>95%
- Cat. No.:76102-632
Specifications
About this item
Fibronectins are adhesive glycoproteins and are found both as cell surface proteins and in plasma.
- Suitable for cell culture
- Presentation: White Solid
- Soluble in water (1 mg/mL at 37 °C).
Human fibronectin (HFN) is suitable for use as an attachment factor in the propagation of cells in vitro when used to coat cell culture surfaces, including plasticware, glassware, and microcarrier beads. It is used in cell attachment and proliferation assays on human endothelial cells, human keratinocytes and human dermal fibroblasts.
HFN is purified from Cohn fraction I of human plasma by a modification of the procedure. This material has been pasteurized to reduce the possibility of viral contamination, and filtered through a 0.22 micron filter prior to lyophilization.
Reconstitution: Reconstitute in physiological buffer or urea for best results. After reconstitution, store in convenient aliquots at -20 °C for up to one year. Avoid repeated freeze/thaw cycles.
HFN is purified from Cohn fraction I of human plasma by a modification of the procedure. This material has been pasteurized to reduce the possibility of viral contamination, and filtered through a 0.22 micron filter prior to lyophilization.