Order Entry
United States
ContactUsLinkComponent
Genomic DNA, Mouse, EMD Millipore
Genomic DNA, Mouse, EMD Millipore
Catalog # EM69239-3
Supplier:  MilliporeSigma
CAS Number:  
Genomic DNA, Mouse, EMD Millipore
Catalog # EM69239-3
Supplier:  MilliporeSigma
Supplier Number:  69239-3
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Mouse Genomic DNA
  • Size:
    100 μg
  • Storage Conditions:
    Store at 2 – 8 °C
  • Cat. No.:
    EM69239-3
  • Supplier no.:
    69239-3

Specifications

About this item

Genomic DNAs are highly purified preparations of high molecular weight genomic DNA from a variety of organisms.

The isolation protocol minimizes shearing and removes contaminants that inhibit enzymatic procedures. Novagen genomic DNA is qualified for genomic analysis (including PCR) and library construction. At least 90% of the DNA is greater than 100 kbp in size, as analyzed by CHEF gel electrophoresis (> 50 kbp for S. cerevisiae). No inhibition of restriction enzyme activity is observed, and the DNA is free of contaminating RNA and protein.

Delivery: DNA is supplied in 10 mM Tris-HCl, pipet with a wide-bore pipet tip to minimize shearing.