About this item
Lipases are a subclass of the esterases. It catalyzes the hydrolysis of emulsified esters of glycerol and long chain fatty acids.
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Lipases catalyze the hydrolysis of esters in aqueous solutions and the synthesis of esters in non-aqueous solutions. They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate). Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.
Activators: Ca2+ is required for activity [Sr2+ and Mg2+ are less effective activators.] Inhibitors: Versene, Zn2+, Cu2+, Hg2+, iodine, PCMB. DFP does not inhibit. Stabilizers: DFP may be used to stabilize impure preparations containing proteinases in solutions.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Source:Pancreas
- Species:Pig
- Storage Conditions:Store at –0 °C.
- Enzyme Name:Lipase
- Enzyme Activity:≥12000 U/g solid