Orders LinkContactUsLinkComponent
Fluorophos® Cheese Extraction Buffer, Advanced Instruments
Fluorophos® Cheese Extraction Buffer, Advanced Instruments
Fluorophos® Cheese Extraction Buffer, 240 mL
Catalog # 52197-478
Supplier #:  FLA005
Fluorophos® Cheese Extraction Buffer, Advanced Instruments
Catalog # 52197-478
CAS Number:  
Fluorophos® Cheese Extraction Buffer, Advanced Instruments
Fluorophos® Cheese Extraction Buffer, 240 mL
CAS Number:  
Price
$208.98
Log in for availability
Pack of 240
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The fluorophos cheese extraction buffer is used to extract alkaline phosphatase from cheese products.
Documents
Specifications
Specifications
  • Description:
    Fluorophos® Cheese Extraction Buffer
  • Size:
    240 mL