31-Piece Standard and Deep Socket Sets, Wright Tool
6-Point, 0.375" Square
300035-397
: 875-339
Measurements provided in inches.
: Set includes (13) Std. Sockets: 0.25 – 1"; (13) Deep Sockets: 0.25 – 1"; ratchet; 3" and 5" extension; flex handle; universal joint; and metal box.
- Description:6-Point
- Drive size:0.375" Square