Order Entry
United States
ContactUsLinkComponent
Sodium pyruvate
Sodium pyruvate
Supplier:  MP Biomedicals
Certificates
undefined
Sodium pyruvate
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test:
    Passes
  • Assay:
    ≥98.0%
  • Solubility (0.06g/ml):
    Clear and Complete
  • Free Acid:
    ≤0.25%

Specification Test Results

Frequently Bought Together