Order Entry
United States
Orders LinkContactUsLinkComponent
Dexamethasone

Dexamethasone

Glass bottle, amber, 50 mg
Catalog # 76177-524
Irritant
Health
Supplier:   MP Biomedicals
Supplier #:  02199350.6
Dexamethasone
Catalog # 76177-524
Supplier:  MP Biomedicals
CAS Number:  50-02-2
Dexamethasone

Glass bottle, amber, 50 mg

CAS Number:  50-02-2
Price
$125.00
Log in for availability
Each (50mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

An anti-inflammatory glucocorticoid with a range of effects on cell survival, cell signaling and gene expression. Use to study apoptosis, cell signaling pathways and gene expression.
Room Temperature

  • Water Soluble Grade
  • Presentation: White Powder
  • Greater than 55 mg dexamethasone per gram with a balance consisting of 2-hydroxypropyl-β-cyclodextrin. Typically used in cell culture applications.
  • Soluble in water (25 mg/ml - clear to slightly hazy, colorless solution)
Synonyms:  Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
  • CAS Number:50-02-2
  • Formula:C₂₂H₂₉FO₅
  • MDL Number:MFCD00064136
  • Molecular Weight:392.47 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identity Test:
    Passes

Specifications

  • Pack type:
    Glass bottle, amber
  • Size:
    50 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.