Order Entry
United States
ContactUsLinkComponent
Phenol red sodium salt solution 0.5%, dark red solution
Phenol red sodium salt solution 0.5%, dark red solution
Catalog # 76180-674
Supplier:  MP Biomedicals
CAS Number:  
undefined
Phenol red sodium salt solution 0.5%, dark red solution
Catalog # 76180-674
Supplier:  MP Biomedicals
Supplier Number:  091690049
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 mL
  • Cat. No.:
    76180-674

Specifications

Specification Test Results

  • Osmolality:
    24 - 55 mM/kg
  • Sterility Test:
    Passes
  • Functionality Test:
    Passes

Specification Test Results

About this item

 :
 :376.35 g/mol
Storage Temperature:Ambient
CAS Number:34487-61-1
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901