Omnifit® BenchMark™ Microbore Chromatography Columns with Two Fixed Endpieces, Diba Industries
Supplier: Antyila Scientific
Certificates
About this item
Designed for applications requiring the smallest bed volumes.
- Precision borosilicate glass columns provide inertness, visibility, and performance
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Delivery: Delivered with two (0.5 and 2 m length) ¹/₁₆" Ext.Ø PTFE tubing assembly with 1/4-28 UNF PP fitting at one end.