About this item
HindIII restriction enzyme recognizes A^AGCTT sites and cuts best at 37 °C in its own unique buffer
- Superior quality, stringent quality control and industry leading manufacturing process
- Convenient color-coded five buffer systems
- Wide selection of restriction endonuclease specificities
- Includes universal Tango buffer for double-digestions
Restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that sent in DNA preparations.
Exhibit 100% activity in the recommended buffer and reaction conditions.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Compatible buffer:FastDigest Green Buffer or FastDigest Buffer (colorless)
- Overhang type:5' overhang
- Enzyme Name:HindIII