Order Entry
United States
ContactUsLinkComponent
Total Iron LRC Comparator Kit, LaMotte
Total Iron LRC Comparator Kit, LaMotte
Catalog # 66170-284
Supplier:  LaMotte
Small Business Enterprise
Total Iron LRC Comparator Kit, LaMotte
Catalog # 66170-284
Supplier:  LaMotte
Supplier Number:  7787-01
Small Business Enterprise

Specifications

  • Description:
    Total Iron LRC Comparator Bar
  • Cat. No.:
    66170-284
  • Range:
    0.05 - 1.0 ppm
  • Supplier no.:
    7787

Specifications

About this item

This kit tests for Low Iron in water using Iron Reagents and the LRC Comparator

Range/Sensitivity is 0.05, 0.10, 0.20, 0.30, 0.40, 0.60, 0.80, 1.0 ppm Fe.

Delivery: It includes 30 mL Iron Reagent #1, 4.5 g Iron Reagent #2 powder, 1 to 5 mL distilled water ampoule, 3 glass test tubes of 10 mL with caps, 1 plastic spoon 0.15 g and Low Range Comparator.